ENAH0424CA67 Enamine

4-chloro-2,5-difluorobenzene-1-sulfonamide

  • MDL number MFCD02091387
  • Cas Number : 287172-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1611986

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1611986
Availability details for
Quantity
  • Availability
  • Add to Cart