ENAH0424E1C1 Enamine

1-cyclopropyl-3-(trifluoromethyl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD31745779
  • Cas Number : 2172246-65-8
  • Molecular Weight : 220.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1617097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1617097
Availability details for
Quantity
  • Availability
  • Add to Cart