ENAH042578F8 Enamine

methyl 2-[(2R)-piperidin-2-yl]acetate hydrochloride

  • MDL number MFCD11506651
  • Cas Number : 144239-68-9
  • Molecular Weight : 193.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1654389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1654389
Availability details for
Quantity
  • Availability
  • Add to Cart