ENAH042579AB Enamine

5-methylhex-5-enoic acid

  • MDL number MFCD02259906
  • Cas Number : 55170-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1657497

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1657497
Availability details for
Quantity
  • Availability
  • Add to Cart