ENAH042579F9 Enamine

1,5-difluoro-3-iodo-2-nitrobenzene

  • MDL number MFCD28789226
  • Cas Number : 771-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1659327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1659327
Availability details for
Quantity
  • Availability
  • Add to Cart