ENAH04257A5B Enamine

methyl 2-(methylamino)-3-phenylpropanoate hydrochloride

  • MDL number MFCD08458660
  • Cas Number : 16975-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1661027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1661027
Availability details for
Quantity
  • Availability
  • Add to Cart