ENAH04257A7D Enamine

(3-methylpyridin-4-yl)methanol

  • MDL number MFCD04038635
  • Cas Number : 38070-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1653554

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1653554
Availability details for
Quantity
  • Availability
  • Add to Cart