ENAH0425804A Enamine

(2S)-4-amino-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid

  • MDL number MFCD00237017
  • Cas Number : 161420-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1556311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1556311
Availability details for
Quantity
  • Availability
  • Add to Cart