ENAH042584FA Enamine

2-bromo-3-chloro-6-methyl-5-nitropyridine

  • MDL number MFCD28734336
  • Cas Number : 1806051-06-8
  • Molecular Weight : 251.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1590060

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1590060
Availability details for
Quantity
  • Availability
  • Add to Cart