ENAH04258645 Enamine

(2R)-1-fluoropropan-2-amine hydrochloride

  • MDL number MFCD30188513
  • Cas Number : 273734-17-1
  • Molecular Weight : 113.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1587667

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1587667
Availability details for
Quantity
  • Availability
  • Add to Cart