ENAH04258653 Enamine

1-(ethylsulfanyl)propane-1-thione

  • MDL number MFCD16038346
  • Cas Number : 998-79-8
  • Molecular Weight : 134.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1588515

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1588515
Availability details for
Quantity
  • Availability
  • Add to Cart