ENAH049F1C31 Enamine

2-cyclopropyl-2,2-difluoroacetic acid

  • MDL number MFCD26728228
  • Cas Number : 1557661-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-171267

Safety Information

signalword Warning
pcodes P305+P351+P338, P337+P313,P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P312,P330,P332+P313,P362,P403+P233,P405,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-171267
pictograms GHS07
hstatements H302,H315,H320,H335
Availability details for
Quantity
  • Availability
  • Add to Cart