ENAH049F1C44 Enamine

tert-butyl 4-(2-oxo-2,3-dihydro-1H-1,3-benzodiazol-1-yl)piperidine-1-carboxylate

  • MDL number MFCD05244849
  • Cas Number : 87120-81-8
  • Molecular Weight : 317.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-102042

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102042
Availability details for
Quantity
  • Availability
  • Add to Cart