ENAH049F1C52 Enamine

1,3-dimethyl-2-[(2-oxopropyl)sulfanyl]-1H-imidazol-3-ium chloride

  • MDL number MFCD00898949
  • Cas Number : 135273-74-4
  • Molecular Weight : 220.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Cl-
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-177572

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177572
Availability details for
Quantity
  • Availability
  • Add to Cart