ENAH049F1C80 Enamine

4-methyl-1,3-oxazole-2-carbaldehyde

  • MDL number MFCD10700246
  • Cas Number : 159015-07-3
  • Molecular Weight : 111.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-52
Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-194448

Safety Information

signalword Warning
pcodes P210,P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-194448
pictograms GHS07
hstatements H227,H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart