ENAH049F1D05 Enamine

[ethynyl(phenyl)phosphoroso]benzene

  • MDL number MFCD31691605
  • Cas Number : 6104-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57-58
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99838

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P273,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-99838
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335,H412
Availability details for
Quantity
  • Availability
  • Add to Cart