ENAH049F1D7F Enamine

tert-butyl 2-cyanomorpholine-4-carboxylate

  • MDL number MFCD18250325
  • Cas Number : 1211592-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-211561

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-211561
Availability details for
Quantity
  • Availability
  • Add to Cart