ENAH049F1E4B Enamine

4-chloro-1,2-dihydro-1,8-naphthyridin-2-one

  • MDL number MFCD24556140
  • Cas Number : 59514-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-301935

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-301935
Availability details for
Quantity
  • Availability
  • Add to Cart