ENAH049F1E9A Enamine

5-(tributylstannyl)-1,2-thiazole

  • MDL number MFCD31616326
  • Cas Number : 1416853-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366889
Availability details for
Quantity
  • Availability
  • Add to Cart