ENAH049F21A2 Enamine

6-oxa-1-azaspiro[3.5]nonane hydrochloride

  • MDL number MFCD28987403
  • Cas Number : 1956324-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1605153

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1605153
Availability details for
Quantity
  • Availability
  • Add to Cart