ENAH049F285E Enamine

6-bromo-8-methoxyimidazo[1,2-a]pyridine

  • MDL number MFCD23708758
  • Cas Number : 1427424-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1693641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1693641
Availability details for
Quantity
  • Availability
  • Add to Cart