ENAH049F2917 Enamine

(1S)-4-chloro-2,3-dihydro-1H-inden-1-amine hydrochloride

  • MDL number MFCD26585389
  • Cas Number : 2109874-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1694209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1694209
Availability details for
Quantity
  • Availability
  • Add to Cart