ENAH049F2D92 Enamine

2-bromo-5-(difluoromethyl)-4-methylpyridine

  • MDL number MFCD25477783
  • Cas Number : 1805019-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1696772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1696772
Availability details for
Quantity
  • Availability
  • Add to Cart