ENAH049F2DED Enamine

5-(benzyloxy)-2-{[(benzyloxy)carbonyl]amino}-5-oxopentanoic acid

  • MDL number MFCD00276402
  • Cas Number : 254980-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1699436

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1699436
Availability details for
Quantity
  • Availability
  • Add to Cart