ENAH049F2EA5 Enamine

tert-butyl (1S,3S,5S)-3-(aminomethyl)-2-azabicyclo[3.1.0]hexane-2-carboxylate

  • MDL number MFCD28501263
  • Cas Number : 1038509-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1696842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1696842
Availability details for
Quantity
  • Availability
  • Add to Cart