ENAH049F2EFC Enamine

1,3-dihydrospiro[indene-2,4'-piperidine]-3-one hydrochloride

  • MDL number MFCD18711589
  • Cas Number : 185525-49-9
  • Molecular Weight : 237.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1700263

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1700263
Availability details for
Quantity
  • Availability
  • Add to Cart