ENAH0CD45894 Enamine

6-chloro-5-nitro-1H-indazole

  • MDL number MFCD08690109
  • Cas Number : 101420-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-412701

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-412701
Availability details for
Quantity
  • Availability
  • Add to Cart