ENAH0CD458EF Enamine

5-methylquinoline

  • MDL number MFCD01685457
  • Cas Number : 7661-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-124556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124556
Availability details for
Quantity
  • Availability
  • Add to Cart