ENAH0CD459C2 Enamine

(3S)-4-cyclohexyl-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid

  • MDL number MFCD08276242
  • Cas Number : 1016276-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1659250

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1659250
Availability details for
Quantity
  • Availability
  • Add to Cart