ENAH0CD459F0 Enamine

4-(hydroxymethyl)bicyclo[2.2.2]octane-1-carboxylic acid

  • MDL number MFCD20640208
  • Cas Number : 828-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-198571

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-198571
Availability details for
Quantity
  • Availability
  • Add to Cart