ENAH0CD45A6C Enamine

1-(2,4-difluoro-3-hydroxyphenyl)ethan-1-one

  • MDL number MFCD16619378
  • Cas Number : 951163-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1698353

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1698353
Availability details for
Quantity
  • Availability
  • Add to Cart