ENAH0CD45AD9 Enamine

5-(4-chlorophenyl)-1-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD08144097
  • Cas Number : 957513-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-206761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-206761
Availability details for
Quantity
  • Availability
  • Add to Cart