ENAH0CD45EA4 Enamine

7-bromo-1H,2H,3H-pyrido[2,3-b][1,4]thiazine

  • MDL number MFCD28348668
  • Cas Number : 1593552-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1704722

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1704722
Availability details for
Quantity
  • Availability
  • Add to Cart