ENAH0CD45EC1 Enamine

2-chloro-4H-thieno[3,2-b]pyrrole-5-carboxylic acid

  • MDL number MFCD11109753
  • Cas Number : 332099-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1703098

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1703098
Availability details for
Quantity
  • Availability
  • Add to Cart