ENAH0CD4605C Enamine

1-(2,4-dichlorophenyl)-2,2,2-trifluoroethan-1-one

  • MDL number MFCD02260844
  • Cas Number : 92736-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1704122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1704122
Availability details for
Quantity
  • Availability
  • Add to Cart