ENAH0CD464F2 Enamine

2-{[(tert-butoxy)carbonyl](3,3,3-trifluoropropyl)amino}acetic acid

  • MDL number MFCD31665357
  • Cas Number : 2219378-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1589622

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1589622
Availability details for
Quantity
  • Availability
  • Add to Cart