ENAH0CD465CA Enamine

dimethyl[(morpholin-3-yl)methyl]amine

  • MDL number MFCD12546268
  • Cas Number : 128454-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1708832

Safety Information

signalword Danger
pcodes P210,P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P370+P378,P403+P233,P403+P235,P405,P501
hcodes C,Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-1708832
pictograms GHS05,GHS07
hstatements H227,H302,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart