ENAH11DCF6B9 Enamine

5-({[(tert-butoxy)carbonyl]amino}methyl)pyridine-2-carboxylic acid

  • MDL number MFCD24047018
  • Cas Number : 864353-57-1
  • Molecular Weight : 252.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-153051

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-153051
Availability details for
Quantity
  • Availability
  • Add to Cart