ENAH11DCF6D4 Enamine

2,2,2-trifluoro-N-(3-methylpiperidin-4-yl)acetamide

  • MDL number MFCD24647588
  • Cas Number : 1862972-61-9
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-173786

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173786
Availability details for
Quantity
  • Availability
  • Add to Cart