ENAH11DCF763 Enamine

2-amino-5-iodo-1,4-dihydropyrimidin-4-one

  • MDL number MFCD30553794
  • Cas Number : 3993-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96198

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-96198
Availability details for
Quantity
  • Availability
  • Add to Cart