ENAH11DCFDEE Enamine

2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-methoxybenzoic acid

  • MDL number MFCD02682215
  • Cas Number : 332121-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-650166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-650166
Availability details for
Quantity
  • Availability
  • Add to Cart