ENAH11DCFDFF Enamine

methyl 4-bromo-2-chloro-1,3-benzothiazole-6-carboxylate

  • MDL number MFCD28967994
  • Cas Number : 1403763-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-658745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-658745
Availability details for
Quantity
  • Availability
  • Add to Cart