ENAH11DCFEAF Enamine

lithium(1+) ion 2-fluoro-5-methylpyridine-3-sulfinate

  • MDL number MFCD31665826
  • Cas Number : 2228434-15-7
  • Molecular Weight : 181.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-753194

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-753194
Availability details for
Quantity
  • Availability
  • Add to Cart