ENAH11DD0134 Enamine

2-oxo-1-(propan-2-yl)-1,2-dihydropyridine-4-carboxylic acid

  • MDL number MFCD26096892
  • Cas Number : 1203544-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-747118

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-747118
Availability details for
Quantity
  • Availability
  • Add to Cart