ENAH11DD0226 Enamine

2-(5-cyclopropyl-1H-1,2,3,4-tetrazol-1-yl)acetic acid

  • MDL number MFCD19282239
  • Cas Number : 1267117-83-8
  • Molecular Weight : 168.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-872798

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-872798
Availability details for
Quantity
  • Availability
  • Add to Cart