ENAH11DD05A9 Enamine

3-[(pyrrolidin-2-yl)methyl]pyridine

  • MDL number MFCD02684095
  • Cas Number : 106366-28-3
  • Molecular Weight : 162.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1238584

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-1238584
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart