ENAH11DD0B92 Enamine

3-fluoro-N-(prop-2-yn-1-yl)aniline

  • MDL number MFCD16661634
  • Cas Number : 436849-26-2
  • Molecular Weight : 149.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1257670

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1257670
Availability details for
Quantity
  • Availability
  • Add to Cart