ENAH11DD100D Enamine

5-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]oxolane-3-carboxylic acid

  • MDL number MFCD31665829
  • Cas Number : 2228434-32-8
  • Molecular Weight : 367.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1696296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1696296
Availability details for
Quantity
  • Availability
  • Add to Cart