ENAH11DD1032 Enamine

tert-butyl N-{[3-(aminomethyl)phenyl]methyl}-N-methylcarbamate

  • MDL number MFCD08689908
  • Cas Number : 954238-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1662309

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1662309
Availability details for
Quantity
  • Availability
  • Add to Cart