ENAH11DD12E5 Enamine

2-chloro-1,3-benzothiazol-6-ol

  • MDL number MFCD00459764
  • Cas Number : 2591-16-4
  • Molecular Weight : 185.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1695559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1695559
Availability details for
Quantity
  • Availability
  • Add to Cart